MilliporeSigma
  • The effect of low dose rate on metabolomic response to radiation in mice.
The effect of low dose rate on metabolomic response to radiation in mice.
Radiation and environmental biophysics (2014-07-23)
Maryam Goudarzi, Tytus D Mak, Congju Chen, Lubomir B Smilenov, David J Brenner, Albert J Fornace
ABSTRACT
Metabolomics has been shown to have utility in assessing responses to exposure by ionizing radiation (IR) in easily accessible biofluids such as urine. Most studies to date from our laboratory and others have employed γ-irradiation at relatively high dose rates (HDR), but many environmental exposure scenarios will probably be at relatively low dose rates (LDR). There are well-documented differences in the biologic responses to LDR compared to HDR, so an important question is to assess LDR effects at the metabolomics level. Our study took advantage of a modern mass spectrometry approach in exploring the effects of dose rate on the urinary excretion levels of metabolites 2 days after IR in mice. A wide variety of statistical tools were employed to further focus on metabolites, which showed responses to LDR IR exposure (0.00309 Gy/min) distinguishable from those of HDR. From a total of 709 detected spectral features, more than 100 were determined to be statistically significant when comparing urine from mice irradiated with 1.1 or 4.45 Gy to that of sham-irradiated mice 2 days post-exposure. The results of this study show that LDR and HDR exposures perturb many of the same pathways such as TCA cycle and fatty acid metabolism, which also have been implicated in our previous IR studies. However, it is important to note that dose rate did affect the levels of particular metabolites. Differences in urinary excretion levels of such metabolites could potentially be used to assess an individual's exposure in a radiobiological event and thus would have utility for both triage and injury assessment.
Product Description
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
5-Hydroxy-L-tryptophan, powder
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)
Sigma-Aldrich
5-Hydroxy-L-tryptophan, 98% (calc. on dried substance)
Sigma-Aldrich
N-Methyl-2-pyridone, ≥99%
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
4-Nitrobenzoic acid, 98%
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Indole, ≥99%, FG
Supelco
Riboflavin (B2), analytical standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard