MilliporeSigma
Analyzing macroautophagy in hepatocytes and the liver.
Methods in enzymology (2009-02-17)
Wen-Xing Ding, Xiao-Ming Yin
ABSTRACT
Mammalian autophagy has been well characterized in the liver and in hepatocytes. Autophagy plays important roles in the normal physiology of the liver and in the pathogenesis of several liver diseases. This chapter will discuss the commonly used methods for analysis of autophagy in hepatocytes and in the liver.
Sigma-Aldrich
Williams′ Medium E, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
SAFC
Williams′ Medium E, With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Williams′ Medium E, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture