MilliporeSigma
  • Binary and ternary atomic layers built from carbon, boron, and nitrogen.
Binary and ternary atomic layers built from carbon, boron, and nitrogen.
Advanced materials (Deerfield Beach, Fla.) (2012-07-14)
Li Song, Zheng Liu, Arava Leela Mohana Reddy, Narayanan Tharangattu Narayanan, Jaime Taha-Tijerina, Juan Peng, Guanhui Gao, Jun Lou, Robert Vajtai, Pulickel M Ajayan
ABSTRACT
Two-dimensional (2D) atomic layers derived from bulk layered materials are very interesting from both scientific and application viewpoints, as evidenced from the story of graphene. Atomic layers of several such materials such as hexagonal boron nitride (h-BN) and dichalcogenides are examples that complement graphene. The observed unconventional properties of graphene has triggered interest in doping the hexagonal honeycomb lattice of graphene with atoms such as boron (B) and nitrogen (N) to obtain new layered structures. Individual atomic layers containing B, C, and N of various compositions conform to several stable phases in the three-component phase diagram of B-C-N. Additionally, stacking layers built from C and BN allows for the engineering of new van-der-Waals stacked materials with novel properties. In this paper, the synthesis, characterization, and properties of atomically thin layers, containing B, C, and N, as well as vertically assembled graphene/h-BN stacks are reviewed. The electrical, mechanical, and optical properties of graphene, h-BN, and their hybrid structure are also discussed along with the applications of such materials.
Sigma-Aldrich
Boron, crystalline, −60 mesh, 99% trace metals basis
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Boron, monofilament, 20m, diameter 0.2mm
Boron, monofilament, 200m, diameter 0.1mm
Boron, monofilament, 20m, diameter 0.1mm
Boron, monofilament, 10m, diameter 0.2mm
Boron, monofilament, 10m, diameter 0.1mm
Boron, monofilament, 50m, diameter 0.1mm
Boron, monofilament, 5m, diameter 0.1mm
Boron, monofilament, 50m, diameter 0.2mm
Boron, monofilament, 5m, diameter 0.2mm