MilliporeSigma
  • [Cost-effectiveness of the addition of acarbose to the treatment of patients with type-2 diabetes in Spain].
[Cost-effectiveness of the addition of acarbose to the treatment of patients with type-2 diabetes in Spain].
Gaceta sanitaria (2007-04-11)
Carme Piñol, Stephane Roze, William Valentine, Thomas Evers
ABSTRACT
To assess the cost-effectiveness of the addition of acarbose to existing treatment in patients with type 2 diabetes mellitus (DM2) in Spain. The CORE Diabetes Model (a published and validated computer simulation model) was used to project long-term clinical and cost outcomes in DM2. Transition probabilities and risk adjustments were derived from published sources. Treatment effects and baseline cohort characteristics were based on a meta-analysis. Direct costs were retrieved from published sources and projected over patient lifetimes from the perspective of the Spanish National Health Service. Costs and clinical benefits were discounted at 3% per year. Sensitivity analyses were performed. Acarbose treatment was associated with improved life expectancy (0.23 years) and quality-adjusted life years (QALY) (0.21 years). Direct costs were on average euro 468 per patient more expensive with acarbose than with placebo. The incremental cost-effectiveness ratios were euro 2,002 per life year gained and euro 2,199 per QALY gained. An acceptability curve showed that with a willingness to pay euro 20,000, which is generally accepted to represent very good value for money, acarbose treatment was associated with a 93.5% probability of being cost-effective. This long-term economic study showed that the addition of acarbose to existing therapy for DM2 was associated with improvements in life expectancy and QALYs in these patients.
Product Description
Acarbose for identification, European Pharmacopoeia (EP) Reference Standard
Acarbose for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acarbose, ≥95% (HPLC)
Supelco
Acarbose, Pharmaceutical Secondary Standard; Certified Reference Material
Acarbose, European Pharmacopoeia (EP) Reference Standard