MilliporeSigma
  • New chiral 4-substituted 2-cyanoethyl-oxazolines: synthesis and assessment of some biological activities.
New chiral 4-substituted 2-cyanoethyl-oxazolines: synthesis and assessment of some biological activities.
Chemico-biological interactions (2014-04-15)
Rym Hassani, Yakdhane Kacem, Hedi Ben Mansour, Hamed Ben Ammar, Béchir Ben Hassine
ABSTRACT
This paper describes the synthesis of new enantiomerically pure 2-cyanoethyl-oxazolines in one step starting from a wide range of amino alcohols and 4-ethoxy-4-iminobutanenitrile with high to good yields (73-96%) via an appropriate procedure which can be used for a selective synthesis of mono-oxazolines. A simple operation as well as a practical separation is additional eco-friendly attributes of this method. All the synthesized compounds were identified and characterized with their physicochemical features and their spectral data ((1)H NMR, (13)C NMR and TOFMS ES(+)). Among the prepared mono-oxazolines, the mono-oxazoline (3a) [3-[(4S)-4-benzyl-4,5-dihydro-1,3-oxazol-2-yl] propanenitrile] was tested to detect some biological activities. This compound was studied in vitro given the various types of pharmacological properties characterizing these compounds such as antioxidant, antimicrobial and analgesic activities. The antioxidant activity and mechanism of (3a) were identified using various in vitro antioxidant assays including 1,1-diphenyl-2-picryl-hydrazyl (DPPH), and superoxide anion radicals (O2(-)) scavenging activity. In addition, compared to Quercetin, the tested synthetic product reveals a relatively-strong antiradical activity towards the DPPH (activity percentage of 81.22%) free radicals and significantly decreased the reactive oxygen species such as (O2(-)) formation evaluated by the non-enzymatic (nitroblue tetrazolium/riboflavine) and the enzymatic (xanthine/xanthine oxidase) systems. Related activity values were, respectively, 66% and 60.30%. The oxazoline (3a) showed a high ability to reduce the O2(-) generation and proved to be a very potent radical scavenger. On the other hand, the analgesic property of the 3[(4S)-benzyl-4,5-dihydro-1,3-oxazol-2-yl] propanenitrile (3a) was demonstrated. The subcutaneous administration of (3a) produced a significant reduction in the number of abdominal constrictions amounting to 73.81% in the acetic acid writhing test in mice. In addition to these advances, the oxazoline (3a) has been investigated as an antimicrobial agent. Our results showed that this molecule exhibited various levels of antibacterial effect against all the tested bacterial strains.
Product Description
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
SAFC
L-Valine
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
(R)-(−)-2-Amino-1-butanol, 98%
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Supelco
Chlorotrimethylsilane, for GC derivatization, LiChropur, ≥99.0% (GC)
USP
L-Leucine, United States Pharmacopeia (USP) Reference Standard