MilliporeSigma
  • Diallyl disulfide suppresses proliferation and induces apoptosis in human gastric cancer through Wnt-1 signaling pathway by up-regulation of miR-200b and miR-22.
Diallyl disulfide suppresses proliferation and induces apoptosis in human gastric cancer through Wnt-1 signaling pathway by up-regulation of miR-200b and miR-22.
Cancer letters (2013-07-16)
Hailin Tang, Yanan Kong, Jiaoli Guo, Yi Tang, Xinhua Xie, Lu Yang, Qi Su, Xiaoming Xie
ABSTRACT
The purpose of this study was to identify a mechanism related to miRNA pathway which plays a role in the anti-tumor effects of Diallyl disulfide. Alterations in miRNA expression were observed in Diallyl disulfide-treated MGC-803 cells, including up-regulation of miR-200b and miR-22 expression. Furthermore, Wnt-1 was identified as a target of both miR-200b and miR-22. MiR-200b and miR-22 not only synergistically inhibited gastric cancer growth, but also enhanced the antitumor effect of Diallyl disulfide both in vitro and in vivo. It indicated that miR-200b and miR-22 may serve as potential gene therapy and enhance Diallyl disulfide antitumor effects.
Sigma-Aldrich
Allyl disulfide, technical grade, 80%
Sigma-Aldrich
Allyl disulfide, ≥80%, FG