MilliporeSigma
  • Synthesis of an Isoindoline-Annulated, Tricyclic Sultam Library via Microwave-Assisted, Continuous-Flow Organic Synthesis (MACOS).
Synthesis of an Isoindoline-Annulated, Tricyclic Sultam Library via Microwave-Assisted, Continuous-Flow Organic Synthesis (MACOS).
Synthesis (2012-01-01)
Farman Ullah, Qin Zang, Salim Javed, Patrick Porubsky, Benjamin Neuenswander, Gerald H Lushington, Paul R Hanson, Michael G Organ
ABSTRACT
A microwave-assisted, continuous-flow organic synthesis (MACOS) protocol for the synthesis of an isoindoline-annulat-ed, tricyclic sultam library, utilizing a Heck-aza-Michael (HaM) strategy, is reported. This sequence involves a Heck reaction on vi-nylsulfonamides with batch microwave heating followed by a one-pot, sequential intramolecular aza-Michael cyclization/Boc-deprot-ection using MACOS. Subsequent cyclization with either 1,1'-carbonyldiimidazole or chloromethyl pivalate using MACOS provided an array of tricyclic sultams. This efficient three-step protocol requires only a few hours to produce the target sultams starting from simple starting materials. Using this strategy, a 38-member library of isoindoline-annulated sultams was generated in good to excellent overall yields (53-87%).
Sigma-Aldrich
Chloromethyl pivalate, 97%