Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

141186

Sigma-Aldrich

Chloromethyl pivalate

97%

Synonym(s):

POM-Cl, Pivaloyloxymethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOOCH2Cl
CAS Number:
Molecular Weight:
150.60
Beilstein:
1560838
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

146-148 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C(=O)OCCl

InChI

1S/C6H11ClO2/c1-6(2,3)5(8)9-4-7/h4H2,1-3H3

InChI key

GGRHYQCXXYLUTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloromethyl pivalate reacts with sodium salt of sulbactam to yield sulbactam pivoxil. It undergoes acylation reaction with 9-(2-phosphonylmethoxyethyl)adenine (PMEA) to yield bis(pivaloyloxymethyl) PMEA.

Application

Chloromethyl pivalate was used in the synthesis of pivaloyloxy methyl ester of ofloxacin as prodrug. It was used as the reagent during the synthesis of an isoindoline-annulated, tricyclic sultam library via microwave-assisted, continuous-flow organic synthesis.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Maeda et al.
Biological & pharmaceutical bulletin, 16(6), 594-599 (1993-06-01)
We newly synthesized a pivaloyloxymethyl ester of ofloxacin (OFLX-PVM) as prodrug in order to avoid the chelate formation between new quinolone and metal cations such as Al3+, Mg2+, Ca2+, or Fe2+ in the gastrointestinal tract. This compound was rapidly hydrolyzed
L S Changov et al.
Farmaco (Societa chimica italiana : 1989), 55(2), 134-135 (2000-04-27)
Sulbactam pivoxil, a prodrug of the beta-lactamase inhibitor sulbactam, was prepared in high yield by reacting the sodium salt of sulbactam with chloromethyl pivalate in a polar solvent, then diluting the reaction mixture with water and isolating the product by
J E Starrett et al.
Antiviral research, 19(3), 267-273 (1992-09-11)
9-(2-Phosphonylmethoxyethyl)adenine (PMEA; 1) was acylated with chloromethyl pivalate to afford bis(pivaloyloxymethyl) PMEA (2). The ester prodrug demonstrated enhanced in vitro potency against HSV-2 greater than 150-fold higher than the parent compound. The antiviral activity of 2 was 50-fold better than
Farman Ullah et al.
Synthesis, 44(16), doi:10-doi:10 (2012-01-01)
A microwave-assisted, continuous-flow organic synthesis (MACOS) protocol for the synthesis of an isoindoline-annulat-ed, tricyclic sultam library, utilizing a Heck-aza-Michael (HaM) strategy, is reported. This sequence involves a Heck reaction on vi-nylsulfonamides with batch microwave heating followed by a one-pot, sequential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service