MilliporeSigma
  • gamma-Glutamylglutamine and taurine concentrations are decreased in the cerebrospinal fluid of drug-naive patients with schizophrenic disorders.
gamma-Glutamylglutamine and taurine concentrations are decreased in the cerebrospinal fluid of drug-naive patients with schizophrenic disorders.
Journal of neurochemistry (1995-12-01)
K Q Do, C J Lauer, W Schreiber, M Zollinger, U Gutteck-Amsler, M Cuénod, F Holsboer
ABSTRACT
HPLC and gas chromatography-mass spectrometry analyses of 18 amino acids, N-acetylaspartate, N-acetylaspartylglutamate, and 5-hydroxyindoleacetic acid, derived from serotonin, and homovanillic acid, derived from dopamine, were performed in CSF collected from a group of patients with schizophrenia who either had been drug free for at least 1 year (n = 5) or were drug naive for psychotropic drugs (n = 21) and in 15 control subjects. Significant differences were found only for taurine (15% lower in the patients) and isoleucine (7% higher). A number of unidentified substances were detected, one of which proved to be markedly reduced (16%) among the schizophrenic patients. Liquid chromatography-mass spectrometry with continuous flow-fast atom bombardment interface allowed us to identify this substance as gamma-glutamylglutamine. The decreased level of gamma-glutamylglutamine may reflect a deficiency in the gamma-glutamyltransferase system, a system probably involved in glutamate uptake, or a deficiency in glutamine, an important precursor of releasable glutamate. Although glutamate was nonsignificantly reduced in the patients, it was one of the five substances (including gamma-glutamylglutamine) that were necessary for the best discrimination between the schizophrenic patients and the controls. These findings support the notion that the glutamatergic system is affected in schizophrenic disorders. In addition, they underscore the need to apply rigid bioanalytical techniques and use drug-naive patients to gain in-depth information on the pathophysiology of brain disorders such as schizophrenia.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
N-Acetyl-L-aspartic acid, ≥99.0% (T)
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
Glycine
SAFC
L-Threonine
SAFC
L-Tyrosine
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, ≥99%
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Alanine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland