MilliporeSigma
  • Quantification of 3-deazaneplanocin A, a novel epigenetic anticancer agent, in rat biosamples by hydrophilic interaction liquid chromatography-tandem mass spectrometric detection.
Quantification of 3-deazaneplanocin A, a novel epigenetic anticancer agent, in rat biosamples by hydrophilic interaction liquid chromatography-tandem mass spectrometric detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-01-05)
Feng Sun, Jasmine Chiat-Ling Ong, Qiang Yu, Eli Chan
ABSTRACT
A sensitive and selective LC-MS/MS based bioanalytical method was developed and validated for the quantification of 3-deazaneplanocin A (DZNep), a novel epigenetic anti-tumor drug candidate, in Sprague-Dawley (SD) rat biosamples (plasma, urine, feces and tissue samples). The method comprises a phenylboronic acid (PBA)-containing solid phase extraction procedure, serving for binding and clean-up of DZNep in rat biosamples spiked with tubercidin (as internal standard). The analytes were separated on an Agilent hydrophilic interaction chromatography (HILIC) column. LC-MS/MS in positive ion mode was used to perform multiple reaction monitoring at m/z of 263/135 and 267/135 for DZNep and tubercidin, respectively. The limit of quantification (LOQ) of DZNep in rat biosamples was 20 ng/mL. The data of intra-day and inter-day accuracy were within 15% of nominal concentration while the precision (relative standard deviation) less than 10% for all biosamples. The extraction recoveries for DZNep and tubercidin were consistent and reproducible (around 80%) and the matrix effects were negligible (around 10% suppression) in all biosamples. This method was demonstrated to be applicable for pharmacokinetic studies of DZNep in SD rats.
Sigma-Aldrich
Tubercidin, from Streptomyces tubercidicus, ~95%