Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

T0642

Sigma-Aldrich

Tubercidin

from Streptomyces tubercidicus, ~95%

Synonym(s):

7-Deazaadenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N4O4
CAS Number:
Molecular Weight:
266.25
Beilstein:
38498
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

biological source

Streptomyces tubercidicus

Assay

~95%

form

powder

color

off-white

antibiotic activity spectrum

fungi
parasites
viruses

Mode of action

DNA synthesis | interferes

SMILES string

O[C@H]1[C@@H](O)[C@H](N2C=CC3=C2N=CN=C3N)O[C@@H]1CO

InChI

1S/C11H14N4O4/c12-9-5-1-2-15(10(5)14-4-13-9)11-8(18)7(17)6(3-16)19-11/h1-2,4,6-8,11,16-18H,3H2,(H2,12,13,14)/t6-,7-,8-,11-/m1/s1

InChI key

HDZZVAMISRMYHH-KCGFPETGSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: nucleoside

Application

It is used to study tubercidin′s mechanism of toxicity, tubercidin resistance and is used as a selection agent. Tubercidin is used to study RNAi silencing. It is used to study the role of transmethylation in chemotaxis of eukaryotic cells.

Biochem/physiol Actions

Tubercidin is a toxic adenosine analog with antiviral, antitrypanosomal, and antifungal functions. It inhibits multiple metabolic processes, including RNA processing, nucleic acid synthesis, protein synthesis, and methylation of tRNA through intracellular incorporation into nucleic acids. Tubercidin acts as a plant antifungal, inhibits mammalian SAH hydrolase (SAHH), and blocks purine biosynthesis in Candida famata. Tubercidin inhibits glycolysis in Trypanosoma brucei. Tubercidin inhibits both chemotaxis and chemotaxis-dependent cell streaming of Dictyostelium, and chemotaxis of neutrophils at concentrations that have little effect on cell viability.
Toxic adenosine analog with antiviral, antitrypanosomal, and antifungal functions. Mode of action: Inhibits multiple metabolic processes, including RNA processing, nucleic acid synthesis, protein synthesis, and methylation of tRNA through intracellular incorporation into nucleic acids. Tubercidin acts as a plant antifungal, inhibits mammalian SAH hydrolase (SAHH), and blocks purine biosynthesis in Candida famata.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aurelie Bourderioux et al.
Journal of medicinal chemistry, 54(15), 5498-5507 (2011-06-30)
A series of 7-aryl- and 7-hetaryl-7-deazaadenosines was prepared by the cross-coupling reactions of unprotected or protected 7-iodo-7-deazaadenosines with (het)arylboronic acids, stannanes, or zinc halides. Nucleosides bearing 5-membered heterocycles at the position 7 exerted potent in vitro antiproliferative effects against a
Misaki Matsui et al.
Oncogenesis, 9(6), 60-60 (2020-06-17)
The nucleus of mammalian cells is compartmentalized by nuclear bodies such as nuclear speckles, however, involvement of nuclear bodies, especially nuclear speckles, in DNA repair has not been actively investigated. Here, our focused screen for nuclear speckle factors involved in
David B Olsen et al.
Antimicrobial agents and chemotherapy, 55(2), 937-939 (2010-12-01)
Efforts to develop novel, interferon-sparing therapies for treatment of chronic hepatitis C (HCV) infection are contingent on the ability of combination therapies consisting of direct antiviral inhibitors to achieve a sustained virologic response. This work demonstrates a proof of concept
Marco Singer et al.
Journal of the American Chemical Society, 132(24), 8372-8377 (2010-05-21)
Photochromic nucleosides were designed that combine the structural features and molecular recognition properties of nucleic acids with the light-sensitivity of diarylethenes. Target compounds 1a-c consist of a 7-deazaadenosine unit that is linked to a thiophene as the second aryl functionality
Feng Sun et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(3-4), 285-290 (2011-01-05)
A sensitive and selective LC-MS/MS based bioanalytical method was developed and validated for the quantification of 3-deazaneplanocin A (DZNep), a novel epigenetic anti-tumor drug candidate, in Sprague-Dawley (SD) rat biosamples (plasma, urine, feces and tissue samples). The method comprises a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service