MilliporeSigma
  • Comparison of most probable number and pour plate procedures for isolation and enumeration of sulphite-reducing Clostridium spores and group D faecal streptococci from oysters.
Comparison of most probable number and pour plate procedures for isolation and enumeration of sulphite-reducing Clostridium spores and group D faecal streptococci from oysters.
The Journal of applied bacteriology (1987-05-01)
T J Easterbrook, P A West
ABSTRACT
A comparative study of methods to enumerate sulphite-reducing Clostridium spores and Group D faecal streptococci in oysters demonstrated that pour plate solid agar techniques gave higher counts than liquid broth most probable number procedures. Reinforced clostridial broth with supplements to detect sulphite reduction was compared with pour plates of egg yolk-free tryptose sulphite cycloserine agar incubated at 37 degrees C for 24 h. Azide dextrose broth was compared with pour plates using Slanetz and Bartley (SB) agar or KF-streptococcus agar at 37 degrees C. Most probable number procedures used for both groups of organisms gave excessive numbers of improbable tube combinations. For enumeration of Group D faecal streptococci, a pour plate technique using SB agar incubated at 37 degrees C for 48 h is recommended.
Product Description
Millipore
Enterococcus Selective Agar, suitable for microbiology, NutriSelect® Basic