MilliporeSigma
Efficient management of fruit pests by pheromone nanogels.
Scientific reports (2013-02-19)
Deepa Bhagat, Suman K Samanta, Santanu Bhattacharya
ABSTRACT
Environment-friendly management of fruit flies involving pheromones is useful in reducing the undesirable pest populations responsible for decreasing the yield and the crop quality. A nanogel has been prepared from a pheromone, methyl eugenol (ME) using a low-molecular mass gelator. This was very stable at open ambient conditions and slowed down the evaporation of pheromone significantly. This enabled its easy handling and transportation without refrigeration, and reduction in the frequency of pheromone recharging in the orchard. Notably the involvement of the nano-gelled pheromone brought about an effective management of Bactrocera dorsalis, a prevalent harmful pest for a number of fruits including guava. Thus a simple, practical and low cost green chemical approach is developed that has a significant potential for crop protection, long lasting residual activity, excellent efficacy and favorable safety profiles. This makes the present invention well-suited for pest management in a variety of crops.
Product Description
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
4-Allyl-1,2-dimethoxybenzene, 99%
Sigma-Aldrich
Methyl eugenol, ≥98%, FCC
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Supelco
Methyl eugenol, analytical standard
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material