Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1269

Supelco

Eugenol

Eugenol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Methoxy-4-(2-propenyl)phenol, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2C=CHCH2)C6H3-2-(OCH3)OH
CAS Number:
Molecular Weight:
164.20
Beilstein:
1366759
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. E5000000
traceable to USP 1268965

API family

eugenol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.541 (lit.)

bp

254 °C (lit.)

mp

−12-−10 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(CC=C)ccc1O

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3,5-7,11H,1,4H2,2H3

InChI key

RRAFCDWBNXTKKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Eugenol may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations[1][2][3] by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0005 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Static headspace gas chromatography of (semi-) volatile drugs in pharmaceuticals for topical use.
Sitaramaraju Y, et al.
Journal of Pharmaceutical and Biomedical Analysis, 47(4-5), 834-840 (2008)
Validated RP-HPLC method to estimate eugenol from commercial formulations like Caturjata Churna, Lavangadi Vati, Jatiphaladi Churna, Sitopaladi Churna and clove oil.
Saran S, et al.
Journal of Pharmacy Research, 6(1), 53-60 (2013)
Quantitative HPTLC analysis of the eugenol content of leaf powder and a capsule formulation of Ocimum sanctum.
Lalla J, et al.
JPC-Journal of Planar Chromatography-Modern TLC, 20(2), 135-138 (2007)
Bhavini Shah et al.
International journal of food microbiology, 161(1), 53-59 (2012-12-25)
There has been great interest in intervention strategies based on plant essential oils to control pathogens such as Escherichia coli O157:H7 and Listeria monocytogenes (Lm). However, the poor solubility of essential oils in water makes it difficult to disperse evenly
Hyang Nam et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 55, 106-112 (2013-01-15)
The oxidative damage of lipid, protein and DNA is known to be involved in chronic inflammation as well as metastasis. It has been highlighted for searching natural compounds without toxicity to prevent development of these diseases. Thus, it was investigated

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service