MilliporeSigma
  • A549 lung cell line activity of biosynthesized silver nanoparticles using Albizia adianthifolia leaf.
A549 lung cell line activity of biosynthesized silver nanoparticles using Albizia adianthifolia leaf.
Colloids and surfaces. B, Biointerfaces (2013-01-29)
R M Gengan, K Anand, A Phulukdaree, A Chuturgoon
ABSTRACT
Stable AgNPs were formed in vitro by reacting AgNO3 (aq) solution with the aqueous plant leaf extract. UV-vis revealed the surface plasmon resonance λmax at 448 nm and the absorbance steadily increased in intensity as a function of reaction time. Transmission electron microscope (TEM) and XRD studies were used to characterize the AgNPs; the size was 4-35 nm. Dynamic light scattering (DLS) was used as supporting evidence to determine hydrodynamic size and zeta potential recorded as 80.27 nm and -24.7 mV, respectively. FT-IR spectra suggest that AgNPs are capped with protein molecules and other water soluble phytocompounds such as saponins and glycosides which also behave as stabilizing agents; TEM images indicate a visible layer surrounding the AgNPs. Prominent absorption bands at 3380 and 1642 cm(-1) are assigned to alcohol and carbonyl groups, respectively. (1)H NMR of the neat aqueous plant extract indicates presence of a complex mixture of compounds; however the chemical shift at δ 6.0-8.0 and 1.0-4.0 ppm indicates the presence of few aromatic but abundant aliphatic compounds, respectively. Toxicity of AgNPs on lung cancer cells (A549) and normal healthy peripheral lymphocytes (PLs) at 10 μg/ml and 50 μg/ml was assessed using the MTT, ATP and lactate dehydrogenase assays. Viability data for A549 cells showed a 21% (10 μg/ml) and 73% (50 μg/ml) cell viability after 6h exposure to AgNPs compared to 117% (10 μg/ml) and 109% (50 μg/ml) cell viability of normal peripheral lymphocytes. Lactate dehydrogenase was only significantly altered at 50 μg/ml AgNPs treated cells from 2.43±0.04 units to 0.77±0.04 units.
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Ag, 1000 ppm Ag
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 0.0282 M
Sigma-Aldrich
Silver nitrate solution, 0.05 M