MilliporeSigma
  • 'One-pot' synthesis of dihydrobenzo[4,5][1,3]oxazino[2,3-a] isoquinolines via a silver(I)-catalyzed cascade approach.
'One-pot' synthesis of dihydrobenzo[4,5][1,3]oxazino[2,3-a] isoquinolines via a silver(I)-catalyzed cascade approach.
Molecules (Basel, Switzerland) (2013-01-25)
Baifeng Jiang, Yu Zhou, Qingya Kong, Hualiang Jiang, Hong Liu, Jian Li
ABSTRACT
An efficient approach for the synthesis of biologically interesting fused tetracyclic isoquinolines in high yields and with a broad substrate scope has been developed. The strategy features an AgNO(3) catalyzed 'one-pot' cascade process involving formation of two new C-N bonds and one new C-O bond.
Sigma-Aldrich
Silver standard solution, suitable for atomic absorption spectrometry, 1 mg/mL Ag, 1000 ppm Ag
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate solution, 0.0282 M
Sigma-Aldrich
Silver nitrate solution, 0.05 M
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O