MilliporeSigma
  • Cis-trans amide bond rotamers in β-peptoids and peptoids: evaluation of stereoelectronic effects in backbone and side chains.
Cis-trans amide bond rotamers in β-peptoids and peptoids: evaluation of stereoelectronic effects in backbone and side chains.
Journal of the American Chemical Society (2013-01-25)
Jonas S Laursen, Jens Engel-Andreasen, Peter Fristrup, Pernille Harris, Christian A Olsen
ABSTRACT
Non-natural peptide analogs have significant potential for the development of new materials and pharmacologically active ligands. One such architecture, the β-peptoids (N-alkyl-β-alanines), has found use in a variety of biologically active compounds but has been sparsely studied with respect to folding propensity. Thus, we here report an investigation of the effect of structural variations on the cis-trans amide bond rotamer equilibria in a selection of monomer model systems. In addition to various side chain effects, which correlated well with previous studies of α-peptoids, we present the synthesis and investigation of cis-trans isomerism in the first examples of peptoids and β-peptoids containing thioamide bonds as well as trifluoroacetylated peptoids and β-peptoids. These systems revealed an increase in the preference for cis-amides as compared to their parent compounds and thus provide novel strategies for affecting the folding of peptoid constructs. By using NMR spectroscopy, X-ray crystallographic analysis, and density functional theory calculations, we present evidence for the presence of thioamide-aromatic interactions through C(sp(2))-H···S(amide) hydrogen bonding, which stabilize certain peptoid conformations.
Sigma-Aldrich
Acrylamide, ≥98.0%
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered