Skip to Content
MilliporeSigma
All Photos(1)

Documents

01-1300

Sigma-Aldrich

Acrylamide

≥98.0%

Synonym(s):

2-Propenamide, Acrylic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCONH2
CAS Number:
Molecular Weight:
71.08
Beilstein:
605349
MDL number:
UNSPSC Code:
41105305
PubChem Substance ID:

vapor density

2.45 (vs air)

vapor pressure

0.03 mmHg ( 40 °C)

Assay

≥98.0%

form

solid

availability

available only in Japan

bp

125 °C/25 mmHg (lit.)

mp

82-86 °C (lit.)

SMILES string

NC(=O)C=C

InChI

1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)

InChI key

HRPVXLWXLXDGHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

for electrophoresis

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

280.4 °F

Flash Point(C)

138 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rusty L Montgomery et al.
EMBO molecular medicine, 6(10), 1347-1356 (2014-09-23)
Over the last decade, great enthusiasm has evolved for microRNA (miRNA) therapeutics. Part of the excitement stems from the fact that a miRNA often regulates numerous related mRNAs. As such, modulation of a single miRNA allows for parallel regulation of
Franco Pedreschi et al.
Journal of the science of food and agriculture, 94(1), 9-20 (2013-08-14)
Acrylamide (AA) is known as a neurotoxin in humans and it is classified as a probable human carcinogen by the International Agency of Research on Cancer. AA is produced as by-product of the Maillard reaction in starchy foods processed at
Arianna Rath et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(39), 15668-15673 (2013-09-11)
SDS/PAGE is universally used in biochemistry, cell biology, and immunology to resolve minute protein amounts readily from tissue and cell extracts. Although molecular weights of water-soluble proteins are reliably determined from their SDS/PAGE mobility, most helical membrane proteins, which comprise
Chun-Hua Lu et al.
Nano letters, 13(3), 1298-1302 (2013-02-21)
Copolymer chains consisting of acrylamide units and guanine (G)-containing oligonucleotide-tethered acrylamide units undergo, in the presence of K(+) ions, cross-linking by G-quadruplexes to yield a hydrogel. The hydrogel is dissociated upon addition of 18-crown-6 ether that traps the K(+) ions.
Jonas S Laursen et al.
Journal of the American Chemical Society, 135(7), 2835-2844 (2013-01-25)
Non-natural peptide analogs have significant potential for the development of new materials and pharmacologically active ligands. One such architecture, the β-peptoids (N-alkyl-β-alanines), has found use in a variety of biologically active compounds but has been sparsely studied with respect to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service