MilliporeSigma
  • Brain efflux index to investigate the influence of active efflux on brain distribution of pemetrexed and methotrexate.
Brain efflux index to investigate the influence of active efflux on brain distribution of pemetrexed and methotrexate.
Drug metabolism and disposition: the biological fate of chemicals (2013-01-09)
Li Li, Sagar Agarwal, William F Elmquist
ABSTRACT
Antifolates, in particular methotrexate (MTX), have been widely used in the treatment of primary and secondary tumors of the central nervous system (CNS). Pemetrexed (PMX) is a novel antifolate that also exhibits potent antitumor activity against CNS malignancies. Studies have shown that brain distribution of both antifolates is significantly restricted, possible due to active efflux transport at the blood-brain barrier (BBB). This study characterizes the brain-to-blood transport of PMX and MTX and examines the role of several efflux transporters in brain distribution of the antifolates by use of the intracerebral microinjection technique (brain efflux index). The results from this study show that both PMX and MTX undergo saturable efflux transport across the BBB, with elimination half-lives of approximately 39 minutes and 29 minutes, respectively. Of the various efflux transporters this study investigated, multidrug resistance-associated protein 2 (Mrp2) does not play an important role in the brain distribution of the two antifolate drugs. Interestingly, breast-cancer resistance protein (Bcrp) makes a significant contribution to the brain elimination of MTX but not PMX. In addition, the brain-to-blood transport of both antifolates was inhibited by probenecid and benzylpenicillin, suggesting the involvement of organic anion transporters in the efflux of these compounds from the brain, with organic anion transporter 3 (Oat3) being a possibility. Our results suggest that one of the underlying mechanisms behind the limited brain distribution of PMX and MTX is active efflux transport processes at the BBB, including a benzylpenicillin-sensitive transport system and/or the active transporter Bcrp.
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Probenecid, European Pharmacopoeia (EP) Reference Standard
Supelco
Penicillin G potassium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Penicillin G potassium salt
Sigma-Aldrich
Probenecid
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%
Sigma-Aldrich
Penicillin G potassium salt, powder, BioReagent, suitable for cell culture