MilliporeSigma
Reactions of latent prints exposed to blood.
Forensic science international (2012-11-28)
Nicole Praska, Glenn Langenburg
ABSTRACT
We explored whether an undeveloped latent print (fingermark) exposed to blood and later developed by enhancement with blood reagents such as amido black (AB) or leucocrystal violet (LCV) could appear as a genuine blood mark. We examined three different experimental conditions. In Experiment I, fingermark residue only was tested, as a control to confirm that fingermark residue alone does not react with the blood reagents AB and LCV. Experiment II investigated whether latent fingermarks exposed to blood dilutions could be treated with AB or LCV and subsequently appear as a genuine blood mark enhanced with AB or LCV. Experiment III tested whether latent fingermarks exposed to whole blood could be processed with AB or LCV and subsequently appear as a genuine blood mark enhanced with AB or LCV. The present study found that indeed, fingermark residue alone does not react with the blood reagents AB and LCV. In Experiment II, an interaction occurred between the fingermark residue and the diluted blood that caused the ridges to appear a red color. In the present study, this interaction is called a faux blood mark. While the faux blood mark phenomenon occurred most often following exposure to diluted blood, it did not occur consistently, and a predictable pattern could not be established. However, the reaction occurred more frequently following extended fingermark residue drying times. Faux blood marks are distinguishable from genuine blood marks prior to enhancement with blood reagents. Following treatment with blood reagents, it became increasingly difficult to determine whether the enhanced mark was a genuine blood print or a latent fingermark exposed to diluted blood. Latent fingermarks exposed to whole blood often resulted in a void prior to enhancement, but following treatment with blood reagents, were difficult to distinguish from a genuine blood mark enhanced with blood reagents.
Sigma-Aldrich
Gram-Hucker′s stain solution (I)
Sigma-Aldrich
Amido Black Staining Solution 2X, electrophoresis reagent
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Gentian Violet solution
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, JIS special grade
Sigma-Aldrich
Crystal Violet, SAJ first grade
Sigma-Aldrich
Crystal Violet, S. No.: 785
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Naphthol Blue Black, Dye content 80 %
Sigma-Aldrich
Naphthol Blue Black, Dye content ~50 %
Sigma-Aldrich
Naphthol Blue Black, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution