MilliporeSigma
  • Exploring the properties of small molecule protein binding via molecular simulations: the TRSH-p53 core domain complex.
Exploring the properties of small molecule protein binding via molecular simulations: the TRSH-p53 core domain complex.
Molecular bioSystems (2012-08-01)
Thomas S Hofer, Wilfred F van Gunsteren
ABSTRACT
Molecular dynamics simulations have been performed to investigate the binding of tris(hydroxymethyl)-aminomethane to the surface of the core domain of the mouse cellular tumor antigen p53 employing the GROMOS and 53A6 force field parameter sets. A close investigation of the crystal structure reported by Ho et al. revealed that the protonated form is bound to the protein, i.e. a tris(hydroxymethyl)-methylammonium ion (TRSH). Molecular Dynamics (MD) simulations indicate that the p53 protein gains stability upon binding the ligand. In addition to MD simulations of the p53 protein with and without the TRSH compound, thermodynamic integration was utilised to estimate the free enthalpy of binding of the TRSH-p53 complex, which was estimated to be -49 and -54 kJ mol(-1) utilising the and 53A6 force fields, respectively.
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine-13C hydrochloride, 99 atom % 13C
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Sigma-Aldrich
Methylamine hydrochloride, ≥98%