MilliporeSigma
  • Investigations on the binding of human hemoglobin with orange I and orange II.
Investigations on the binding of human hemoglobin with orange I and orange II.
Journal of photochemistry and photobiology. B, Biology (2012-05-30)
Yan-Qing Wang, Hong-Mei Zhang
ABSTRACT
The interactions between human hemoglobin and orange I (or orange II) were investigated by UV/vis absorption, circular dichroism, fluorescence spectra techniques, and molecular modeling method. Orange I and orange II effectively quenched the intrinsic fluorescence of human hemoglobin by static quenching. The processes of the binding orange I and orange II on human hemoglobin were spontaneous molecular interaction procedure with hydrogen bonds, van der Waals force, hydrophobic and electrostatic interactions according to van't Hoff equation and molecular modeling. There is a single class of binding site of orange I (orange II) in human hemoglobin and the molecular modeling study shows that orange I and orange II are dipped into α(2) chain. The results of CD, synchronous fluorescence and three-dimensional fluorescence spectra indicated a small loss of α-helical secondary structure of human hemoglobin induced by orange I and orange II.
Product Description
Supelco
Orange II sodium salt, analytical standard
Sigma-Aldrich
Orange II sodium salt, (Certified by the Biological Stain Commission), Dye content ≥85 %