MilliporeSigma
  • Particle size and magnetic properties dependence on growth temperature for rapid mixed co-precipitated magnetite nanoparticles.
Particle size and magnetic properties dependence on growth temperature for rapid mixed co-precipitated magnetite nanoparticles.
Nanotechnology (2012-03-22)
Mei Fang, Valter Ström, Richard T Olsson, Lyubov Belova, K V Rao
ABSTRACT
Magnetite nanoparticles have been prepared by co-precipitation using a custom-designed jet mixer to achieve rapid mixing (RM) of reactants in a timescale of milliseconds. The quick and stable nucleation obtained allows control of the particle size and size distribution via a more defined growth process. Nanoparticles of different sizes were prepared by controlling the processing temperature in the first few seconds post-mixing. The average size of the nanoparticles investigated using a Tecnai transmission electron microscope is found to increase with the temperature from 3.8 nm at 1 ± 1 °C to 10.9 nm for particles grown at 95 ± 1 °C. The temperature dependence of the size distribution follows the same trend and is explained in terms of Ostwald ripening of the magnetite nanoparticles during the co-precipitation of Fe(2+) and Fe(3+). The magnetic properties were studied by monitoring the blocking temperature via both DC and AC techniques. Strikingly, the obtained RM particles maintain the high magnetization (as high as ∼88 A m(2) kg(-1) at 500 kA m(-1)) while the coercivity is as low as ∼12 A m(-1) with the expected temperature dependence. Besides, by adding a drop of tetramethylammonium hydroxide, aqueous ferrofluids with long term stability are obtained, suggesting their suitability for applications in ferrofluid technology and biomedicine.
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium chloride solution, Molecular Biology
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O