MilliporeSigma
  • Verbascoside, isoverbascoside, and their derivatives recovered from olive mill wastewater as possible food antioxidants.
Verbascoside, isoverbascoside, and their derivatives recovered from olive mill wastewater as possible food antioxidants.
Journal of agricultural and food chemistry (2012-01-25)
Angela Cardinali, Sandra Pati, Fiorenza Minervini, Isabella D'Antuono, Vito Linsalata, Vincenzo Lattanzio
ABSTRACT
Olive oil processing industries generate substantial quantities of phenolic-rich byproducts, which could be valuable natural sources of antioxidants. This work is focused on the recovery and structural characterization of antioxidant compounds from olive mill wastewater (OMWW), a polluting byproduct of the olive oil production process. Phenolics were extracted from the waste material using a membrane technology coupled to low-pressure gel filtration chromatography on a Sephadex LH-20. The LH-20 fraction was, in turn, characterized for its phenolic composition by HPLC-DAD-MS/MS analyses. Verbascoside, isoverbascoside, β-hydroxyverbascoside, β-hydroxyisoverbascoside, and various oxidized phenolics were identified. Uptake of verbascoside, purified from the LH-20 fraction, by HT-29 cells, an established model system for studying drug transport properties, was also assayed. Finally, the antioxidant activities of the LH-20 fraction and verbascoside were characterized by two different techniques. Individual verbascoside was more active as a scavenger of reactive oxygen species and as a chemopreventive agent protecting low-density lipoproteins from oxidative damage than the LH-20 fraction.
Sigma-Aldrich
Verbascoside, ≥99% (HPLC)
Verbascoside, primary reference standard