Skip to Content
MilliporeSigma

V4015

Verbascoside

≥99% (HPLC)

Synonym(s):

Acetoside, Kusaginin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H36O15
CAS Number:
Molecular Weight:
624.59
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

room temp

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCc3ccc(O)c(O)c3)O[C@H](CO)[C@H]2OC(=O)\C=C\c4ccc(O)c(O)c4)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C29H36O15/c1-13-22(36)23(37)24(38)29(41-13)44-27-25(39)28(40-9-8-15-3-6-17(32)19(34)11-15)42-20(12-30)26(27)43-21(35)7-4-14-2-5-16(31)18(33)10-14/h2-7,10-11,13,20,22-34,36-39H,8-9,12H2,1H3/b7-4+/t13-,20+,22-,23+,24+,25+,26+,27+,28+,29-/m0/s1

InChI key

FBSKJMQYURKNSU-ZLSOWSIRSA-N

General description

Verbascoside, also known as acteoside, is a hydrophilic plant compound first isolated from mullein (Verbascum sinuatum L.). The chemical structure of verbascoside comprises hydroxyphenylethyl and cinnamic acid moieties bound to a β-glucopyranose via a glycosidic bond.

Application

Verbascoside has been used:
  • as a reference standard for high-performance liquid chromatography (HPLC) analysis
  • as a reference standard for detection of phenolic compounds in olive mill wastewater and pomace by high-performance liquid chromatography (HPLC)-diode-array detector (DAD)-qTOF system
  • to test its anti-viral activity against respiratory syncytial virus (RSV) in HEP2 cells and A549 cells

Biochem/physiol Actions

Phenylethanoid glycoside. Antioxidant and antiproliferative.
Verbascoside has anti-inflammatory and photoprotective properties. It has shown neuroprotective effects on PC12 neuronal cells against neurotoxicity. Verbascoside is proven to be a potent inhibitor of xanthine oxidase (XOD) in in vitro studies.


Still not finding the right product?

Explore all of our products under Verbascoside


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library