MilliporeSigma
  • Organic matter compounds as source indicators and tracers for marine pollution in a western Mediterranean coastal zone.
Organic matter compounds as source indicators and tracers for marine pollution in a western Mediterranean coastal zone.
Environmental science and pollution research international (2011-05-25)
Jalila Amorri, Claude Geffroy-Rodier, Fehmi Boufahja, Ezzeddine Mahmoudi, Patricia Aïssa, Mohamed Ksibi, André Amblès
ABSTRACT
Complex organic compounds found in oil and sediments linked with a particular source (such as algae, bacteria or vascular plants) are defined as biomarkers and are useful dating indicators in organic geochemistry. This paper presents the composition of the organic matter (OM) on marine surface sediments from a degraded Tunisian coast analysed by pyrolysis and gas chromatography-mass spectrometry (GC-MS). High total OM contents (0.3-4.2%) were detected with high levels of saturated linear hydrocarbons. The aliphatic lipids had contributed with up to 11.7% of the total OM, and their distribution had consisted of resolved compounds (n-alkanes and fatty acid (FAs)) and an unresolved complex mixture. Hydrocarbons, primarily n-alkanes, were ranged from 368 to 3,886 μg g(-1). The FAs (674-2,568 μg g(-1)) were dominated by derived primary production, and the short chain FAs (C16 and C18) were the most abundant throughout. The ubiquitous presence of petroleum contamination, mainly from offshore oil exploration, discharge of pollutants from rivers, shipping activities and atmospheric deposition was found in all samples. The Gabès littoral seems to be quite to very polluted near the industrial zone of Ghannouch. The C/H ratio (generally around 5.9), the thermal analysis and GC-MS of n-alkanes and FAs showed that the OM in the studied area was composed of anthropogenic/petrogenic, marine and continental sources. Our study represents an innovative approach to assessing environmental pollution. The evaluation of organic matter by examination of sterols, alkanes and fatty acids allows the identification of source, both anthropogenic and natural.
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium chloride solution, Molecular Biology
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O