MilliporeSigma
  • [Study on the chemical constituents from pine needles of Cedrus deodara].
[Study on the chemical constituents from pine needles of Cedrus deodara].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-06-26)
Jun-min Zhang, Xiao-feng Shi, Chong Li, Bin Fan, Dong-dong Wang, Dong-yan Liu
ABSTRACT
To study the chemical constituents from pine needles of Cedrus deodara. Chemical constituents were isolated by silica gel and Sephadex LH-20 column chromatography. The structures of the isolated compounds were elucidated through spectroscopic analysis. The compounds were identified as 9-hydroxy-dodecanoic acid (I), ethyl laurate (II), ethyl stearate (III), 3beta-hydroxy-oleanolic acid methyl ester (IV), beta-sitosterol (V), shikimic acid (VI), methylconiferin (VII), ferulic acid beta-glucoside (VIII). Compounds I-IV, VI-VIII are isolated and identified from this genus for the first time, compound V is isolated from pine needles of this genus for the first time.
Sigma-Aldrich
Ethyl stearate, ≥99% (capillary GC)
Sigma-Aldrich
Ethyl stearate, ≥97%
Sigma-Aldrich
Ethyl laurate, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl laurate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl dodecanoate, ≥98.0% (GC)