MilliporeSigma
  • Syntheses of novel chiral sulfinamido ligands and their applications in diethylzinc additions to aldehydes.
Syntheses of novel chiral sulfinamido ligands and their applications in diethylzinc additions to aldehydes.
The Journal of organic chemistry (2007-02-10)
Zhiyan Huang, Hongshan Lai, Yong Qin
ABSTRACT
A novel class of sulfinamido alcohol ligands 1-6 was synthesized from (S)-tert-butanesulfinamide. These ligands showed excellent catalytic activities and enantiomeric selectivities in asymmetric additions of diethylzinc to aromatic aldehydes.
Sigma-Aldrich
Diethylzinc, ≥52 wt. % Zn basis
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane
Sigma-Aldrich
Diethylzinc, packaged for use in deposition systems