Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

296112

Sigma-Aldrich

Diethylzinc solution

1.0 M in hexanes

Synonym(s):

Zincdiethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2Zn
CAS Number:
Molecular Weight:
123.51
Beilstein:
3587207
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

1.0 M in hexanes

density

0.726 g/mL at 25 °C

SMILES string

CC[Zn]CC

InChI

1S/2C2H5.Zn/c2*1-2;/h2*1H2,2H3;

InChI key

HQWPLXHWEZZGKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylzinc solution can be used as a reagent to prepare:
  • α-trifluoromethyl ketones from α, β-unsaturated ketones with Rh (I) catalyst.
  • Alkynylzinc reagents which are used in cross-coupling reactions.
  • Chiral secondary alcohols from aromatic and aliphatic aldehydes in presence of camphor-derived β-amino alcohol.
  • 1, 4 conjugate addition product with enones.
  • α-fluoroacrylates from aldehydes and ketones.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective copper-catalyzed conjugate addition using chiral diaminocarbene ligands
Guillen F, et al.
Tetrahedron Asymmetry, 12(15), 2083-2086 (2001)
Novel and general cross-coupling reactions of alkynylzinc reagents and organotellurium compounds
Dabdoub MJ, et al.
Tetrahedron Letters, 41(4), 437-440 (2000)
Asymmetric addition of diethylzinc to aldehydes catalyzed by a camphor-derived β-amino alcohol
Wu Z-L, et al.
Tetrahedron Asymmetry, 20(13), 1556-1560 (2009)
Rhodium-Catalyzed Novel Trifluoromethylation at the α-Position of α, β-Unsaturated Ketones
Sato K, et al.
Organic Letters, 6(23), 4359-4361 (2004)
Organic Syntheses, 83, 177-177 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service