MilliporeSigma
Structure of the human lactate dehydrogenase B gene.
The Biochemical journal (1989-02-01)
T Takeno, S S Li
ABSTRACT
Human genomic clones containing parts of the lactate dehydrogenase B (LDH-B) gene (approx. 25 kb in length) were isolated and characterized. The protein-coding sequence of human LDH-B gene is interrupted by six introns at codons nos. 42-43, 82, 140, 198, 237 and 278-279, and the positions of these introns are homologous to those of LDH-A genes from man and mouse. The 5' non-coding region of human LDH-B gene is interrupted by an intron six nucleotide residues upstream of the ATG translation-initiation site, whereas those of human and mouse LDH-A genes are interrupted at 24 nucleotide residues 5' to the ATG initiation codon. As is the case of LDH-A genes from man and mouse, there is no intron in the 3' non-coding region of human LDH-B gene.
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein