Skip to Content
MilliporeSigma

Skip To

59747

Lactic Dehydrogenase, recombinant from E. coli

≥90 U/mg

Synonym(s):

(S)-Lactate: NAD+ oxidoreductase, L-Lactate Dehydrogenase, Lactate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-617-8
MDL number:
EC Number:
Specific activity:
≥90 U/mg
Recombinant:
expressed in E. coli
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

form

powder

specific activity

≥90 U/mg

storage temp.

−20°C

General description

LDH (lactic dehydrogenase), a glycolytic enzyme, particularly present in skeletal muscle, heart, liver, kidneys, brain, lungs and red blood cells. It has five isoenzyme forms. LDH possess a tetrameric structure.

Application

Lactic Dehydrogenase, recombinant from E. coli has been used:
  • in lactate dehydrogenase (LDH) and malate dehydrogenase 1 (MDH1)assays and cross-linking assays
  • to prepare assay buffer to measure pyruvate kinase (PYK) by coupled assay
  • in in vitro DltC D-alanylation assay

Biochem/physiol Actions

L-lactic dehydrogenase catalyzes the conversion of L-lactate into L-pyruvate while reducing NAD+ to NADH and H+.
Conversion of L-lactate into L-pyruvate is crucial in hypoxic and anaerobic conditions, especially when synthesis of adenosine triphosphate (ATP) by oxidative phosphorylation is interrupted.

Other Notes

One unit corresponds to the amount of enzyme which reduces 1 μmol pyruvate per minute at pH 7.4 and 25°C (NADH as cofactor)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L3916L1378L2625
description

≥90 U/mg

description

1000 units/mL

description

Type X, ammonium sulfate suspension, ≥600 units/mg protein

description

Type III, ammonium sulfate suspension, ≥500 units/mg protein

specific activity

≥90 U/mg

specific activity

1000 units/mL

specific activity

≥600 units/mg protein

specific activity

≥500 units/mg protein

recombinant

expressed in E. coli

recombinant

-

recombinant

-

recombinant

-

form

powder

form

solution

form

ammonium sulfate suspension

form

ammonium sulfate suspension

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–4 of 4 Questions  
  1. Does this enzyme catalyze the reaction of L-lactate to L-pyruvate, with NAD+ -> NADH as cofactors, or does it catalyze the conventional L-pyruvate to L-lactate with NADH -> NAD+?

    1 answer
    1. This enzyme catalyzes the conversion of L-lactate into L-pyruvate while reducing NAD+ to NADH and H+.

      Helpful?

  2. Can you provide more information on the biological source for recombinant LDH 59747? Does the certificate of origin specify whether the recombinant protein is from mice or another source? It seems that the certificate of origin does not offer detailed information to clarify this.

    1 answer
    1. The material is Lactate dehydrogenase from chicken heart, expressed in recombinant microbes.

      Helpful?

  3. I want to know the molecular weight of this protein

    1 answer
    1. This product is a recombinant enzyme from E. coli. The species and tissue of origin is the chicken heart. The molecular weight of this material has not been determined. However, according to Huston et al. 1972, Lactate Dehydrogenase from the bovine heart has a molecular weight of approximately 136,700 daltons.

      Helpful?

  4. What species is Product No. 59747, Lactic Dehydrogenase, recombinant from E. coli?

    1 answer
    1. This product is a recombinant Lactic Dehydrogenase from chicken heart made in E. coli.

      Helpful?

Reviews

No rating value

Active Filters