MilliporeSigma
  • Novel insights into Mycobacterium antigen Ag85 biology and implications in countermeasures for M. tuberculosis.
Novel insights into Mycobacterium antigen Ag85 biology and implications in countermeasures for M. tuberculosis.
Critical reviews in eukaryotic gene expression (2012-11-13)
XieMei Tang, Wanyan Deng, Jianping Xie
ABSTRACT
Tuberculosis remains one of the most prevalent and deadly infectious diseases, largely due to the emergence of multidrug-resistant and extensive drug-resistant Mycobacterium tuberculosis, especially the coinfection with HIV. Mycobacterium Ag85 complex (Ag85A, B, and C), with a carboxylesterase consensus sequence and conserved surface catalysis residues, involves in cell wall biosynthesis and the trigger of the host immune response. The physiological function, structures, distributions, and molecular mechanisms of regulations as well as their implications in novel vaccines and diagnostics against tuberculosis are summarized. Special focus is the regulation underlying the Ag85 expression. This will facilitate in-depth understanding of the role of Ag85 and developing better novel measures against M. tuberculosis infection.
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg