Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352204
EC Number:
232-773-7
NACRES:
NA.54
Specific activity:
≥10 U/mg
Recombinant:
expressed in E. coli
recombinant
expressed in E. coli
form
crystalline, crystals, powder or flakes
specific activity
≥10 U/mg
storage temp.
−20°C
General description
Esterase belongs to the hydrolase superfamily of enzymes.This recombinant esterase contains a C-terminal histidine tag.
Application
Esterase, from Bacillus subtilis, may be used in protein engineering research as well as to study the kinetic resolution of acetates of arylaliphatic tertiary alcohols. Product 96667 is recombinant and expressed in E. Coli (≥10 U/mg).
Biochem/physiol Actions
An esterase is a hydrolase that splits esters into acids and alcohols.
Esterase participates in the stereospecific hydrolysis and production of esters. Esterases, that are obtained from cultured bacteria and fungi has several industrial applications.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Other Notes
1 U corresponds to the amount of enzyme which converts 1 μmol 4-nitrophenyl-L-acetate per minute at pH 7.5 and 30°C.
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
