MilliporeSigma
  • Adrenergic signalling to astrocytes in anterior cingulate cortex contributes to pain-related aversive memory in rats.
Adrenergic signalling to astrocytes in anterior cingulate cortex contributes to pain-related aversive memory in rats.
Communications biology (2023-01-06)
Zafar Iqbal, Zhuogui Lei, Aruna S Ramkrishnan, Shu Liu, Mahadi Hasan, Mastura Akter, Yuk Yan Lam, Ying Li
ABSTRACT
Pain contains both sensory and affective dimensions. We identify the role of norepinephrine in colorectal distention (sub-threshold for acute pain) induced conditioned place avoidance and plasticity gene expression in the anterior cingulate cortex (ACC). Activating locus coeruleus (LC)-projecting ACC neurons facilitates pain-evoked aversive consolidation and memory, while inhibiting LC-projecting ACC neurons reversibly blocks it. Optogenetic activation of ACC astrocytes facilitates aversive behaviour. ACC astrocytic Gi manipulation suppressed aversive behaviour and early plasticity gene expression induced by opto-activation of LC neurons projecting to ACC. Evidences for the critical role of β2AR in ACC astrocytes were provided using AAV encoding β2AR miRNAi to knockdown β2AR in astrocytes. In contrast, opto-activation of ACC astrocytic β2ARs promotes aversion memory. Our findings suggest that projection-specific adrenergic astrocytic signalling in ACC is integral to system-wide neuromodulation in response to visceral stimuli, and plays a key role in mediating pain-related aversion consolidation and memory formation.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-phospho-CREB (Ser133) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Dopamine β Hydroxylase-SAP Antibody, clone 4F10.2, clone 4F10.2, Chemicon®, from mouse
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Clozapine N-oxide hydrochloride, ≥98% (HPLC), Water soluble Clozapine N-oxide
Sigma-Aldrich
Anti-CREB Antibody, clone NL904, rabbit monoclonal, culture supernatant, clone NL904, Upstate®