MilliporeSigma
HDLs extract lipophilic drugs from cells.
Journal of cell science (2022-01-05)
Adi Zheng, Gilles Dubuis, Maria Georgieva, Carla Susana Mendes Ferreira, Marc Serulla, Maria Del Carmen Conde Rubio, Evgeniya Trofimenko, Thomas Mercier, Laurent Decosterd, Christian Widmann
ABSTRACT
High-density lipoproteins (HDLs) prevent cell death induced by a variety of cytotoxic drugs. The underlying mechanisms are however still poorly understood. Here, we present evidence that HDLs efficiently protect cells against thapsigargin (TG), a sarco/endoplasmic reticulum (ER) Ca2+-ATPase (SERCA) inhibitor, by extracting the drug from cells. Drug efflux could also be triggered to some extent by low-density lipoproteins and serum. HDLs did not reverse the non-lethal mild ER stress response induced by low TG concentrations or by SERCA knockdown, but HDLs inhibited the toxic SERCA-independent effects mediated by high TG concentrations. HDLs could extract other lipophilic compounds, but not hydrophilic substances. This work shows that HDLs utilize their capacity of loading themselves with lipophilic compounds, akin to their ability to extract cellular cholesterol, to reduce the cell content of hydrophobic drugs. This can be beneficial if lipophilic xenobiotics are toxic but may be detrimental to the therapeutic benefit of lipophilic drugs such as glibenclamide.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Roche
Transcriptor Universal cDNA Master, sufficient for 100 reactions, suitable for cDNA synthesis, suitable for RT-PCR, suitable for RT-qPCR