Skip to Content
MilliporeSigma
All Photos(1)

Documents

05893151001

Roche

Transcriptor Universal cDNA Master

sufficient for 100 reactions, suitable for cDNA synthesis, suitable for RT-PCR, suitable for RT-qPCR

Synonym(s):

RT-PCR | cDNA synthesis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106313
NACRES:
NA.55

Quality Level

usage

sufficient for 100 reactions

feature

dNTPs included: no
hotstart: no

manufacturer/tradename

Roche

packaging

kit of for 100 reactions

technique(s)

RT-PCR: suitable
RT-qPCR: suitable
cDNA synthesis: suitable

input

purified RNA

detection method

probe-based

General description

Ready-to-use reaction mix for two-step RT-qPCR on all real-time PCR systems.

The Transcriptor Universal cDNA Master is the convenient solution for time-saving cDNA synthesis for two-step real-time RT-PCR. All reagents required, including random hexamer primer, nucleotides, buffers and enzymes are supplied in just two vials minimizing pipetting. Maximum convenience is achieved by combining the Transcriptor Universal cDNA Master mix with the RealTime ready Cell Lysis Kit, further accelerating real-time PCR by directly using cell lysates. The unique enzyme included in this kit has a broad temperature range and is suitable for high temperature reverse transcription. The Transcriptor Universal cDNA Master produces very high specificity and sensitivity, even with difficult complex RNA templates. Excellent performance and linear response in real-time RT-PCR is obtained for both low and high copy genes. The Transcriptor Universal cDNA Master is compatible with LightCycler® instruments and all other real-time PCR instruments requiring normalization with a reference dye.
Sensitive and accurate RT-PCR assay is a prerequisite for meaningful molecular genetic analyses. Choosing the correct enzyme for reverse transcription and cDNA synthesis is critical for obtaining high yields of high quality full-length cDNA, that is faithful to the original cellular RNA.
We offer a comprehensive selection of Transcriptor reverse transcription products, including:

  • Enzymes with exquisite performance and broad dynamic range in real-time PCR.
  • Enzymes for high temperature reverse transcription to synthesize cDNA from RNA with secondary structures.
  • High Fidelity Transcriptor products, with proofreading activity, for applications requiring the highest sequence accuracy.

Application

Reverse transcription is a hallmark of molecular biology. The ability to convert RNA into DNA for qualitative and quantitative RNA analysis using PCR or real-time PCR has revolutionized gene expression analysis. Quantitative reverse transcription polymerase chain reaction (RT-PCR or qRT-PCR) is now the standard for mRNA detection and quantification, sensitive enough to quantify RNA from a single cell.

RT-PCR is also used for the creation of cDNA libraries, molecular cloning by PCR, and sequencing. The Transcriptor Universal cDNA Master is a convenient fast solution for cDNA synthesis and real-time PCR analysis. No matter which PCR instrument you are using, this master mix, supplied in just two vials, brings out the full potential of your qRT-PCR assays.

Features and Benefits

  • Conserve valuable resources with effortless setup.
Just add your RNA. The two vials included in this kit contain all reagents required for reverse transcription, including random hexamer primer, nucleotides, buffers and enzymes
  • Save time with a fast protocol.
The optimized master mix significantly reduces reverse transcription times (10 minutes) and setup times.
  • Maximize convenience by combining this master mix with the Roche RealTime ready Cell Lysis Kit for cell lysates without RNA purification.
  • Power through the most difficult templates.
Transcriptor Reverse Transcriptase′s high thermostability and specially optimized buffer system permit incubation temperatures up to +60°C, accurately transcribing even the most challenging secondary structures.
  • Reliably standardize and quantify your qRT-PCR experiments.
Obtain sensitive reproducible results over a linear dynamic range of at least six orders of magnitude.
  • Increase the sensitivity and specificity of your two-step qRT-PCR.
Produce lower cycle threshold (Ct) values, starting with target amounts as low as 0.1 pg total RNA.

Components

  • Transcriptor Universal Reverse Transcriptase, 20x concentrated
  • Transcriptor Universal Reaction Buffer, 5x concentrated

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

LightCycler is a registered trademark of Roche

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetically unmatched human iPSC and ESC exhibit equivalent gene expression and neuronal differentiation potential
Marei, et al.
Scientific Reports, 7 (2017)
Sex Dependent Hepatoprotective Role of IL 22 Receptor Signaling in Non Alcoholic Fatty Liver Disease Related Fibrosis
Abdelnabi, et al.
Cellular and molecular gastroenterology and hepatology, 14, 1269-1269 (2022)
Adi Zheng et al.
Journal of cell science, 135(5) (2022-01-05)
High-density lipoproteins (HDLs) prevent cell death induced by a variety of cytotoxic drugs. The underlying mechanisms are however still poorly understood. Here, we present evidence that HDLs efficiently protect cells against thapsigargin (TG), a sarco/endoplasmic reticulum (ER) Ca2+-ATPase (SERCA) inhibitor
Monia Souidi et al.
International journal of molecular sciences, 22(2) (2021-01-15)
Numerous protocols of cardiac differentiation have been established by essentially focusing on specific growth factors on human pluripotent stem cell (hPSC) differentiation efficiency. However, the optimal environmental factors to obtain cardiac myocytes in network are still unclear. The mesoderm germ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service