MilliporeSigma
  • Myelin-Independent Therapeutic Potential of Canine Glial-Restricted Progenitors Transplanted in Mouse Model of Dysmyelinating Disease.
Myelin-Independent Therapeutic Potential of Canine Glial-Restricted Progenitors Transplanted in Mouse Model of Dysmyelinating Disease.
Cells (2021-11-28)
Luiza Stanaszek, Malgorzata Majchrzak, Katarzyna Drela, Piotr Rogujski, Joanna Sanford, Michal Fiedorowicz, Magdalena Gewartowska, Malgorzata Frontczak-Baniewicz, Piotr Walczak, Barbara Lukomska, Miroslaw Janowski
ABSTRACT
Dysfunction of glia contributes to the deterioration of the central nervous system in a wide array of neurological disorders, thus global replacement of glia is very attractive. Human glial-restricted precursors (hGRPs) transplanted intraventricularly into neonatal mice extensively migrated and rescued the lifespan in half of the studied mice, whereas mouse GRPs (mGRPs) presented no therapeutic benefit. We studied in the same experimental setting canine GRPs (cGRP) to determine whether their therapeutic potential falls between hGRPs and mGRPs. Additional motivation for the selection of cGRPs was a potential for use in veterinary medicine. cGRPs were extracted from the brain of dog fetuses. The cells were transplanted into the anterior or posterior aspect of the lateral ventricle (LV) of neonatal, immunodeficient, dysmyelinated mice (Mbpshi, Rag2 KO; shiv/rag2). Outcome measures included early cell biodistribution, animal survival and myelination assessed with MRI, immunohistochemistry and electron microscopy. Grafting of cGRP into posterior LV significantly extended animal survival, whereas no benefit was observed after anterior LV transplantation. In contrast, myelination of the corpus callosum was more prominent in anteriorly transplanted animals. The extended survival of animals after transplantation of cGRPs could be explained by the vicinity of the transplant near the brain stem.
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 129-138, clone 1, culture supernatant, clone 1, Chemicon®
Sigma-Aldrich
Anti-CNPase Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-A2B5 Antibody, clone A2B5-105, clone A2B5-105, Chemicon®, from mouse
Sigma-Aldrich
Anti-Galactocerebroside Antibody, clone mGalC, clone mGalC, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig-2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Olig1 Antibody, Chemicon®, from rabbit