MilliporeSigma
  • Miniaturized 3D bone marrow tissue model to assess response to Thrombopoietin-receptor agonists in patients.
Miniaturized 3D bone marrow tissue model to assess response to Thrombopoietin-receptor agonists in patients.
eLife (2021-06-02)
Christian A Di Buduo, Pierre-Alexandre Laurent, Carlo Zaninetti, Larissa Lordier, Paolo M Soprano, Aikaterini Ntai, Serena Barozzi, Alberto La Spada, Ida Biunno, Hana Raslova, James B Bussel, David L Kaplan, Carlo L Balduini, Alessandro Pecci, Alessandra Balduini
ABSTRACT
Thrombocytopenic disorders have been treated with the Thrombopoietin-receptor agonist Eltrombopag. Patients with the same apparent form of thrombocytopenia may respond differently to the treatment. We describe a miniaturized bone marrow tissue model that provides a screening bioreactor for personalized, pre-treatment response prediction to Eltrombopag for individual patients. Using silk fibroin, a 3D bone marrow niche was developed that reproduces platelet biogenesis. Hematopoietic progenitors were isolated from a small amount of peripheral blood of patients with mutations in ANKRD26 and MYH9 genes, who had previously received Eltrombopag. The ex vivo response was strongly correlated with the in vivo platelet response. Induced Pluripotent Stem Cells (iPSCs) from one patient with mutated MYH9 differentiated into functional megakaryocytes that responded to Eltrombopag. Combining patient-derived cells and iPSCs with the 3D bone marrow model technology allows having a reproducible system for studying drug mechanisms and for individualized, pre-treatment selection of effective therapy in Inherited Thrombocytopenias.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Quinacrine dihydrochloride, ≥90%
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
Anti-TRA-1-81 Antibody, clone TRA-1-81, Cy3 conjugate, clone TRA-1-81, from mouse, CY3 conjugate
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-NANOG Antibody, clone 7F7.1, clone 7F7.1, from mouse