MilliporeSigma
  • Developing structure-activity relationships for the prediction of hepatotoxicity.
Developing structure-activity relationships for the prediction of hepatotoxicity.
Chemical research in toxicology (2010-06-18)
Nigel Greene, Lilia Fisk, Russell T Naven, Reine R Note, Mukesh L Patel, Dennis J Pelletier
ABSTRACT
Drug-induced liver injury is a major issue of concern and has led to the withdrawal of a significant number of marketed drugs. An understanding of structure-activity relationships (SARs) of chemicals can make a significant contribution to the identification of potential toxic effects early in the drug development process and aid in avoiding such problems. This process can be supported by the use of existing toxicity data and mechanistic understanding of the biological processes for related compounds. In the published literature, this information is often spread across diverse sources and can be varied and unstructured in quality and content. The current work has explored whether it is feasible to collect and use such data for the development of new SARs for the hepatotoxicity endpoint and expand upon the limited information currently available in this area. Reviews of hepatotoxicity data were used to build a structure-searchable database, which was analyzed to identify chemical classes associated with an adverse effect on the liver. Searches of the published literature were then undertaken to identify additional supporting evidence, and the resulting information was incorporated into the database. This collated information was evaluated and used to determine the scope of the SARs for each class identified. Data for over 1266 chemicals were collected, and SARs for 38 classes were developed. The SARs have been implemented as structural alerts using Derek for Windows (DfW), a knowledge-based expert system, to allow clearly supported and transparent predictions. An evaluation exercise performed using a customized DfW version 10 knowledge base demonstrated an overall concordance of 56% and specificity and sensitivity values of 73% and 46%, respectively. The approach taken demonstrates that SARs for complex endpoints can be derived from the published data for use in the in silico toxicity assessment of new compounds.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Sigma-Aldrich
2-Ethylhexyl 4-methoxycinnamate, 98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Chloroform solution, NMR reference standard, 50% in acetone-d6 (99.9 atom % D), chromium(III) acetylacetonate 0.2 %
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Nicotinic acid, analytical standard
Supelco
Chloroform solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Supelco
L-Ascorbic acid, analytical standard
Supelco
Chloroform solution, NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Chloroform solution, NMR reference standard, 20% in acetone-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Bismuth(III) subsalicylate, 99.9% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Methoxyacetic acid, 98%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Supelco
2-Ethylhexyl 4-methoxycinnamate, analytical standard
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Bromobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%