MilliporeSigma
  • Genome-wide CRISPR Screens Reveal Host Factors Critical for SARS-CoV-2 Infection.
Genome-wide CRISPR Screens Reveal Host Factors Critical for SARS-CoV-2 Infection.
Cell (2020-11-05)
Jin Wei, Mia Madel Alfajaro, Peter C DeWeirdt, Ruth E Hanna, William J Lu-Culligan, Wesley L Cai, Madison S Strine, Shang-Min Zhang, Vincent R Graziano, Cameron O Schmitz, Jennifer S Chen, Madeleine C Mankowski, Renata B Filler, Neal G Ravindra, Victor Gasque, Fernando J de Miguel, Ajinkya Patil, Huacui Chen, Kasopefoluwa Y Oguntuyo, Laura Abriola, Yulia V Surovtseva, Robert C Orchard, Benhur Lee, Brett D Lindenbach, Katerina Politi, David van Dijk, Cigall Kadoch, Matthew D Simon, Qin Yan, John G Doench, Craig B Wilen
ABSTRACT
Identification of host genes essential for severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection may reveal novel therapeutic targets and inform our understanding of coronavirus disease 2019 (COVID-19) pathogenesis. Here we performed genome-wide CRISPR screens in Vero-E6 cells with SARS-CoV-2, Middle East respiratory syndrome CoV (MERS-CoV), bat CoV HKU5 expressing the SARS-CoV-1 spike, and vesicular stomatitis virus (VSV) expressing the SARS-CoV-2 spike. We identified known SARS-CoV-2 host factors, including the receptor ACE2 and protease Cathepsin L. We additionally discovered pro-viral genes and pathways, including HMGB1 and the SWI/SNF chromatin remodeling complex, that are SARS lineage and pan-coronavirus specific, respectively. We show that HMGB1 regulates ACE2 expression and is critical for entry of SARS-CoV-2, SARS-CoV-1, and NL63. We also show that small-molecule antagonists of identified gene products inhibited SARS-CoV-2 infection in monkey and human cells, demonstrating the conserved role of these genetic hits across species. This identifies potential therapeutic targets for SARS-CoV-2 and reveals SARS lineage-specific and pan-CoV host factors that regulate susceptibility to highly pathogenic CoVs.
Sigma-Aldrich
PFI-3, ≥98% (HPLC)
Sigma-Aldrich
SIS3, ≥98% (HPLC), powder
Sigma-Aldrich
MDL 28170, ≥90% (TLC)