Skip to Content
MilliporeSigma

SML0939

PFI-3

≥98% (HPLC)

Synonym(s):

(E)-1-(2-Hydroxyphenyl)-3-((1R,4R)-5-(pyridin-2-yl)-2,5-diazabicyclo[2.2.1]heptan-2-yl)prop-2-en-1-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H19N3O2
CAS Number:
Molecular Weight:
321.37
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(C(/C=C/N2[C@H](C3)CN(C4=NC=CC=C4)[C@H]3C2)=O)C=CC=C1

InChI

1S/C19H19N3O2/c23-17-6-2-1-5-16(17)18(24)8-10-21-12-15-11-14(21)13-22(15)19-7-3-4-9-20-19/h1-10,14-15,23H,11-13H2/b10-8+/t14-,15-/m1/s1

InChI key

INAICWLVUAKEPB-QSTFCLMHSA-N

Biochem/physiol Actions

PFI-3 is a SCG probe for SMARCA 2 and SMARCA 4, members of the large ATP-dependent chromatin remodeling complex SNF/SWI.
SMARCA4 (SWI/SNF related, Matrix associated, Actin dependent Regulator of Chromatin, subfamily A, member 4) is a transcriptional activator and is a component of the large ATP-dependent chromatin remodeling complex SWI/SNF, which is required for transcriptional activation of genes normally repressed by chromatin. SMARC4 (also known as BRG1) and the related protein SMARCA2 (also known as BRM) contain a bromodomain that is structurally similar to the PolyBromo1 (PB1) 5 bromodomain. PFI-3 is a selective chemical probe for SMARCA bromodomains that inhibits SMARCA2, SMARCA4 and PB1(5) bromodomains. For full characterization details, please visit the PFI-3 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

PFI-3 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library