MilliporeSigma
  • The mitochondrial carrier SFXN1 is critical for complex III integrity and cellular metabolism.
The mitochondrial carrier SFXN1 is critical for complex III integrity and cellular metabolism.
Cell reports (2021-03-18)
Michelle Grace Acoba, Ebru S Selen Alpergin, Santosh Renuse, Lucía Fernández-Del-Río, Ya-Wen Lu, Oleh Khalimonchuk, Catherine F Clarke, Akhilesh Pandey, Michael J Wolfgang, Steven M Claypool
ABSTRACT
Mitochondrial carriers (MCs) mediate the passage of small molecules across the inner mitochondrial membrane (IMM), enabling regulated crosstalk between compartmentalized reactions. Despite MCs representing the largest family of solute carriers in mammals, most have not been subjected to a comprehensive investigation, limiting our understanding of their metabolic contributions. Here, we functionally characterize SFXN1, a member of the non-canonical, sideroflexin family. We find that SFXN1, an integral IMM protein with an uneven number of transmembrane domains, is a TIM22 complex substrate. SFXN1 deficiency leads to mitochondrial respiratory chain impairments, most detrimental to complex III (CIII) biogenesis, activity, and assembly, compromising coenzyme Q levels. The CIII dysfunction is independent of one-carbon metabolism, the known primary role for SFXN1 as a mitochondrial serine transporter. Instead, SFXN1 supports CIII function by participating in heme and α-ketoglutarate metabolism. Our findings highlight the multiple ways that SFXN1-based amino acid transport impacts mitochondrial and cellular metabolic efficiency.
Sigma-Aldrich
Anti-SFXN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
4,6-Dioxoheptanoic acid, powder
Sigma-Aldrich
BPTES, ≥95% (HPLC)
Sigma-Aldrich
ALT Activity Assay, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-SFXN3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
sn-Glycerol 3-phosphate lithium salt, ≥95.0% (TLC)
Supelco
1,4-Benzoquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Coenzyme Q9, ≥96.0% (HPLC)
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)
Sigma-Aldrich
n-Dodecyl β-D-maltoside, ≥98% (GC)
Sigma-Aldrich
Protoporphyrin IX, ≥95%
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)
Sigma-Aldrich
Decylubiquinone, ≥97% (HPLC)
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Palmitoyl-L-carnitine chloride, ≥98% (TLC), powder
Deferoxamine mesylate salt, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid
Sigma-Aldrich
Oxalic acid dihydrate, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Dodecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Dimethyl 2-oxoglutarate, 96%