MilliporeSigma
  • Inhibition studies of a beta-carbonic anhydrase from Brucella suis with a series of water soluble glycosyl sulfanilamides.
Inhibition studies of a beta-carbonic anhydrase from Brucella suis with a series of water soluble glycosyl sulfanilamides.
Bioorganic & medicinal chemistry letters (2010-03-10)
Daniela Vullo, Isao Nishimori, Andrea Scozzafava, Stephan Köhler, Jean-Yves Winum, Claudiu T Supuran
ABSTRACT
A beta-carbonic anhydrase (CA, EC 4.2.1.1) from the bacterial pathogen Brucella suis, bsCA 1, has been cloned, purified characterized kinetically and for inhibition with a series of water soluble glycosylated sulfanilamides. bsCA 1 has appreciable activity as catalyst for the hydration of CO(2) to bicarbonate, with a k(cat) of 6.4x10(5) s(-1) and k(cat)/K(m) of 3.9x10(7) M(-1) s(-1). All types of inhibitory activities have been detected, with K(I)s in the range of 8.9-110 nM. The best bsCA 1 inhibitor were the galactose and ribose sulfanilamides, with inhibition constants of 8.9-9.2 nM. Small structural changes in the sugar moiety led to dramatic differences of enzyme inhibitory activity for this series of compounds. One of the tested glycosylsulfonamides and acetazolamide significantly inhibited the growth of the bacteria in cell cultures.
Product Description
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)