MilliporeSigma
  • Dysregulation of intestinal epithelial CFTR-dependent Cl- ion transport and paracellular barrier function drives gastrointestinal symptoms of food-induced anaphylaxis in mice.
Dysregulation of intestinal epithelial CFTR-dependent Cl- ion transport and paracellular barrier function drives gastrointestinal symptoms of food-induced anaphylaxis in mice.
Mucosal immunology (2020-06-25)
Amnah Yamani, David Wu, Richard Ahrens, Lisa Waggoner, Taeko K Noah, Vicky Garcia-Hernandez, Catherine Ptaschinski, Charles A Parkos, Nicholas W Lukacs, Asma Nusrat, Simon P Hogan
ABSTRACT
Food-triggered anaphylaxis can encompass a variety of systemic and intestinal symptoms. Murine-based and clinical studies have revealed a role for histamine and H1R and H2R-pathway in the systemic response; however, the molecular processes that regulate the gastrointestinal (GI) response are not as well defined. In the present study, by utilizing an IgE-mast cell (MC)-dependent experimental model of oral antigen-induced anaphylaxis, we define the intestinal epithelial response during a food-induced anaphylactic reaction. We show that oral allergen-challenge stimulates a rapid dysregulation of intestinal epithelial transcellular and paracellular transport that was associated with the development of secretory diarrhea. Allergen-challenge induced (1) a rapid intestinal epithelial Cftr-dependent Cl- secretory response and (2) paracellular macromolecular leak that was associated with modification in epithelial intercellular junction proteins claudin-1, 2, 3 and 5, E-cadherin and desmosomal cadherins. OVA-induced Cftr-dependent Cl- secretion and junctional protein degradation was rapid occurring and was sustained for 72 h following allergen-challenge. Blockade of both the proteolytic activity and Cl- secretory response was required to alleviate intestinal symptoms of food-induced anaphylaxis. Collectively, these data suggest that the GI symptom of food-induced anaphylactic reaction, secretory diarrhea, is a consequence of CFTR-dependent Cl- secretion and proteolytic activity.
Sigma-Aldrich
CFTR Inhibitor II, GlyH-101, CFTR Inhibitor II, GlyH-101, CAS 328541-79-3, is a cell-permeable potent, selective and reversible open-channel blocker of CFTR (Ki = 4.3 µM in CFTR-expressing FRT cells).
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Claudin 3 antibody produced in rabbit, affinity isolated antibody