Skip to Content
MilliporeSigma

G9545

Anti-GAPDH Antibody

rabbit polyclonal

Synonym(s):

Anti Gapdh, Anti Gapdh Antibody, Anti-Gapdh, GAPDH Antibody - Anti-GAPDH antibody produced in rabbit, Gapdh Antibody, Gapdh Antibody Sigma, Anti-38-kD component, Anti-G3PD, Anti-GAPD, Anti-Glyceraldehyde-3-phosphate dehydrogenase, Anti-OCAS, Anti-OCT1 coactivator in S phase, Anti-p38 component

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IP, WB
Citations:
1546
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution

biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~36 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using mouse NIH3T3 cell lysates, indirect immunofluorescence: 5-10 μg/mL using rat NRK cells, western blot: 0.1-0.2 μg/mL using whole extract of human HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAPDH(2597)
mouse ... Gapdh(14433)
rat ... Gapdh(24383)

General description

The GAPDH (glyceraldehyde-3-phosphate dehydrogenase) gene is mapped to human chromosome 12p13.3 and encodes a tetramer containing identical chains. The enzyme is commonly known as glycolytic enzyme and contains a binding site for NAD+(nicotinamide adenine diphosphate) and glyceraldehyde-3-phosphate. GAPDH contributes to 10-20% of the total cellular protein and is considered to be evolutionarily conserved.

Immunogen

Synthetic peptide corresponding to amino acids of mouse GAPDH, conjugated to KLH via an N-terminal cysteine residue. The corresponding sequence is identical in rat and differs by two amino acids in humans.

Application

Anti-GAPDH antibody produced in rabbit is suitable as a primary antibody for western blotting using:
  • human cancer cell extract
  • extract from muscle tissue from old mice exhibiting sarcopenia
  • trabeculae or samples of the mice heart myocardium
  • mouse embryonic fibroblasts
It is suitable for immunoprecipitation using mouse NIH3T3 cell lysates with 5-10μg, for indirect immunofluorescence using rat NRK cells at a working concentration of 5-10μg/mL. It is also suitable for western blotting at a working concentration of 0.1-0.2μg/mL using a whole extract of human HeLa cells.

Biochem/physiol Actions

Anti-GAPDH recognizes human, mouse, and rat GAPDH.
The enzyme glyceraldehyde-3-phosphate dehydrogenase catalyzes the reversible oxidative phosphorylation of glyceraldehyde-phosphate in the presence of inorganic phosphate and NAD, which is a critical energy-yielding step in carbohydrate metabolism. It binds to several proteins including actin, tubulin, amyloid precursor, polyglutamine peptides, DRPLA (dentatorubral-pallidoluysian atrophy), and huntingtin. GAPDH forms a part of OCA-S, the multicomponent OCT1 (octamer-motif-binding factor) coactivator complex, which is involved in the S phase-dependent histone H2B transcription. This association is responsible for linking H2B transcriptional machinery to cell cycle regulation and to the cellular metabolic state. In response to oxidative stress, GAPDH induces apoptosis.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

For continuous use, store at 2-8 °C for up to one month.
For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library