MilliporeSigma
  • Iron regulatory protein 2 is required for artemether -mediated anti-hepatic fibrosis through ferroptosis pathway.
Iron regulatory protein 2 is required for artemether -mediated anti-hepatic fibrosis through ferroptosis pathway.
Free radical biology & medicine (2020-09-19)
Yujia Li, Chun Jin, Min Shen, Zhenyi Wang, Shanzhong Tan, Anping Chen, Shijun Wang, Jiangjuan Shao, Feng Zhang, Zili Zhang, Shizhong Zheng
ABSTRACT
Currently, the existing treatments have not cured the liver fibrosis thoroughly. Ferroptosis is a newly discovered way of cell death, which is closely related to many diseases. Previous studies have shown that ferroptosis plays an important role in the occurrence and development of liver fibrosis, but the further mechanism remains to be discovered. LX-2 cells were used as the research object, fibrosis activation index was detected by Western blot, PCR and Immunofluorescence, ferroptosis was detected by kits, the binding and interaction between IRP2 (iron regulatory protein 2) and STUB1 (STIP1 homology and U-box containing protein 1) were detected by Immunoprecipitation and ubiquitin test, and IRP2 knockdown mice were constructed by interfering plasmid to verify the results of in vitro experiment. Our research showed that ART (artemether) had a good anti-fibrosis effect in vivo and in vitro, and ferroptosis played an important role in this process. Further studies have found that ART could lead to the accumulation of IRP 2 a in hepatic stellate cell by inhibiting the ubiquitination of it, thus inducing the increase of iron in HSC (hepatic stellate cell), which could product a large number of ROS (reactive oxide species), resulting the occurrence of ferroptosis in cells. Our findings provided an experimental basis for ART to become a drug for the treatment of liver fibrosis. Our results show that IRP2-Iron-ROS axis is necessary for ART to induce ferroptosis in HSC and play an anti-fibrotic effect.
Sigma-Aldrich
Artemether, ≥98% (HPLC)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Carbon-13C tetrachloride, 99 atom % 13C
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%