P26600
2-Phenylindole
technical grade, 95%
Synonym(s):
NSC 15776, Stabilizer I
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Assay
95%
form
powder
bp
250 °C/10 mmHg (lit.)
mp
188-190 °C (lit.)
SMILES string
c1ccc(cc1)-c2cc3ccccc3[nH]2
InChI
1S/C14H11N/c1-2-6-11(7-3-1)14-10-12-8-4-5-9-13(12)15-14/h1-10,15H
InChI key
KLLLJCACIRKBDT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Reactant for preparation of:
Reactant in:
- Oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition
- Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
- Organic light emitting diodes (OLEDs)
- Anti inflammatory agents
- Potential cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors
- Agents for cancer and malaria therapy
- Antifungal and antibacterial agents
- Fluorescent probes
Reactant in:
- Difluorohydroxylation reactions†
- Mannich-type reactions†
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jenan Al-Matouq et al.
Molecular carcinogenesis, 58(9), 1691-1700 (2019-06-27)
Non-melanoma skin cancer frequently results from chronic exposure to ultraviolet (UV) irradiation. UV-induced DNA damage activates cell cycle arrest checkpoints through degradation of the cyclin-dependent kinase activators, the cell division cycle 25 (CDC25) phosphatases. We previously reported increased CDC25A in
Rosemary E McDowell et al.
Investigative ophthalmology & visual science, 57(11), 4762-4771 (2016-09-13)
We studied whether the accumulation of advanced lipoxidation end-products (ALEs) in the diabetic retina is linked to the impairment of lipid aldehyde detoxification mechanisms. Retinas were collected from nondiabetic and diabetic rats and processed for conventional and quantitative RT-PCR (qRT-PCR)
Grace Y Wang et al.
The American journal of surgical pathology, 43(8), 1112-1122 (2019-04-18)
In recent years, a novel small round cell sarcoma harboring EWSR1-NFATC2 translocation with immunomorphologic overlap with Ewing sarcoma (ES), myoepithelial tumors, and extraskeletal myxoid chondrosarcoma has emerged. There has not been a case series devoted to describing its detailed clinicopathologic
Claire Stenhouse et al.
Biology of reproduction, 100(1), 239-252 (2018-08-24)
Inadequate fetal growth cannot be remedied postnatally, leading to severe consequences for neonatal and adult development. It is hypothesized that growth restriction occurs due to inadequate placental vascularization. This study investigated the relationship between porcine fetal size, sex, and placental
Bianca N Cruz et al.
Frontiers in microbiology, 10, 1864-1864 (2019-08-29)
Marine picocyanobacteria are ubiquitous primary producers across the world's oceans, and play a key role in the global carbon cycle. Recent evidence stemming from in situ investigations have shown that picocyanobacteria are able to sink out of the euphotic zone
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service