MilliporeSigma
  • Highly selective ratiometric fluorescent probes for the detection of Fe3+ and its application in living cells.
Highly selective ratiometric fluorescent probes for the detection of Fe3+ and its application in living cells.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2019-11-14)
Chunqing Li, Qi Sun, Qiang Zhao, Xinjian Cheng
ABSTRACT
It's of vital importance to detect heavy metals in environment and living cells. In this work, four near-infrared regions boron dipyrromethene (BODIPY) probes (QBPH, PBPH, QBP and PBP) are constructed based on two BODIPY precursors (QB, PB) for sensing of Fe3+. As expected, these four probes exhibit obvious colorimetric and ratiometric response to Fe3+. In addition, QBP and PBP display highly sensitive and selective performance for detection of Fe3+. More importantly, QBP and PBP are successfully applied to near infrared imaging and detection of Fe3+ in living A549 cells; it indicates that these novel designed probes could be a useful tool for the studies of Fe3+ in living cells.
Sigma-Aldrich
4-Quinolinecarboxaldehyde, 97%
Sigma-Aldrich
2,4-Dimethylpyrrole, 97%
Sigma-Aldrich
3-Pyridinecarboxaldehyde, 98%