MilliporeSigma
  • GluA1 signal peptide determines the spatial assembly of heteromeric AMPA receptors.
GluA1 signal peptide determines the spatial assembly of heteromeric AMPA receptors.
Proceedings of the National Academy of Sciences of the United States of America (2016-09-08)
Xue-Yan He, Yan-Jun Li, Chakrapani Kalyanaraman, Li-Li Qiu, Chen Chen, Qi Xiao, Wen-Xue Liu, Wei Zhang, Jian-Jun Yang, Guiquan Chen, Matthew P Jacobson, Yun Stone Shi
ABSTRACT
AMPA-type glutamate receptors (AMPARs) mediate fast excitatory neurotransmission and predominantly assemble as heterotetramers in the brain. Recently, the crystal structures of homotetrameric GluA2 demonstrated that AMPARs are assembled with two pairs of conformationally distinct subunits, in a dimer of dimers formation. However, the structure of heteromeric AMPARs remains unclear. Guided by the GluA2 structure, we performed cysteine mutant cross-linking experiments in full-length GluA1/A2, aiming to draw the heteromeric AMPAR architecture. We found that the amino-terminal domains determine the first level of heterodimer formation. When the dimers further assemble into tetramers, GluA1 and GluA2 subunits have preferred positions, possessing a 1-2-1-2 spatial assembly. By swapping the critical sequences, we surprisingly found that the spatial assembly pattern is controlled by the excisable signal peptides. Replacements with an unrelated GluK2 signal peptide demonstrated that GluA1 signal peptide plays a critical role in determining the spatial priority. Our study thus uncovers the spatial assembly of an important type of glutamate receptors in the brain and reveals a novel function of signal peptides.
Sigma-Aldrich
Anti-Glutamate Receptor 2 & 3 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Stargazin Antibody, Chemicon®, from rabbit