Skip to Content
MilliporeSigma

AB9876

Anti-Stargazin Antibody

Chemicon®, from rabbit

Synonym(s):

Stargazin-like TARP (transmembrane AMPA receptor regulatory protein)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, IP, WB
Citations:
16
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Cacng2(12300)
rat ... Cacng2(84347)

Immunogen

GST fusion protein from rat stargazin, cytoplasmic domain.

Application

Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
This Anti-Stargazin Antibody is validated for use in IP, WB, IC, IH for the detection of Stargazin.
Western blot: 0.1 μg/mL on Triton soluble fraction of rat brain. Suggested dilution buffer is TBS-T and blocking buffer is 3% skim milk. Suggested incubation time is 3 hours to overnight at room temperature.
Immunohistochemistry: 0.25-0.5 μg/mL on 4% paraformaldehyde fixed tissue. Suggested dilution buffer is PBS-T. Suggested permeabilization method is 0.1% Triton X-100. Suggested incubation time is 2 hours to overnight at room temperature.
Immunocytochemistry
Immunoprecipitation: 1-2 μg. Suggested reaction volume is 200-1000 μL. Suggested total protein concentration is 2 mg/mL. Suggested incubation time is 2 hours at 2-8°C. Suggested capture agent is Protein A.

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

Stargazin [Stargazin-like TARP (transmembrane AMPA receptor regulatory protein)]. By Western blot the antibody recognizes bands at ~35 kDa and ~55 kDa on Triton soluble fraction of rat brain. The antibody reacts dominantly with TARP gamma-8 and weakly with gamma-3 and gamma-4. Stargazin is expressed strongly in the hippocampus and cerebellum.

Physical form

Format: Purified
ImmunoAffinity Purified
Liquid in 130 mM Tris-Cl, 87 mM Glycine, pH 7.4.

Preparation Note

Maintain at -80°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze thaw cycles. The antibody may be kept at 2-8°C for 1-3 weeks after thawing.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library